Skip to Content
Merck
All Photos(1)

Key Documents

40050-U

Supelco

Hexachlorobutadiene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

InChI

1S/C4Cl6/c5-1(3(7)8)2(6)4(9)10

InChI key

RWNKSTSCBHKHTB-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Cord-Ruwisch et al.
Journal of biotechnology, 142(2), 151-156 (2009-05-19)
The microbial reductive dechlorination of chlorinated solvents is a redox reaction in which the chlorinated carbon receives electrons from a suitable electron donor. Hexachloro-1,3-butadiene (HCBD) is a particularly recalcitrant chlorinated solvent. Its slow, cyanocobalamin-dependent biochemical dechlorination by mixed microbial consortia
Edoardo Zanetti et al.
Journal of applied toxicology : JAT, 30(2), 142-150 (2009-09-11)
Little is known concerning the sensitivity of aged rats to xenobiotics inducing kidney damage. To increase this knowledge, the age-dependent response of the kidney to hexachloro-1 : 3-butadiene (HCBD) or potassium dichromate (chromate) was investigated. Rats were treated at different
Arianna Chiusolo et al.
Toxicology letters, 182(1-3), 84-90 (2008-09-23)
Gene expression of regucalcin (Rgn), a calcium-binding protein, was investigated in kidney of male Wistar rats treated with proximal tubule segment-specific nephrotoxicants, namely hexachloro-1:3-butadiene (HCBD), specific for S(3) segment (pars recta) and potassium dichromate (chromate) specific for S(1)-S(2) segments (pars
Aubrey Swain et al.
Journal of applied toxicology : JAT, 32(6), 417-428 (2011-09-10)
Hexachloro-1:3-butadiene (HCBD) causes damage specifically to the renal proximal tubule in rats. In the present study, injury to the nephron of male Hanover Wistar rats was characterized at 24 h following dosing with HCBD in the range 5-90 mg kg⁻¹ to determine the
Chon-Lin Lee et al.
Chemosphere, 58(11), 1503-1516 (2005-02-08)
Forty sediments were analyzed for chlorobenzenes (CBs) and hexachlorobutadiene (HCBD) to investigate their pollution topography along the Kaohsiung coast, Taiwan. Maximum CB concentrations found, varied from 15.4ng/gdw for 1,2,3-TCB to 56.8ng/gdw for 1,2,4-TCB, while higher HCBD concentrations were detected among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service