Skip to Content
Merck
All Photos(1)

Key Documents

40015

Supelco

Chloroethane solution

1000 μg/mL in methanol, analytical standard

Synonym(s):

Ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroethane is clear and colourless gas with a pungent type of odor, which can be commonly used in topical vapocoolant sprays.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This product is now available as a certified reference solution. Please reference CRM40015

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapocoolant Spray Effectiveness on Arterial Puncture Pain: A Randomized Controlled Clinical Trial
Farahmand S, et al.
Acta Medica Iranica, 55, 87-91 (2017)
Suzuko Yamazaki et al.
The journal of physical chemistry. A, 114(15), 5092-5098 (2010-03-27)
The photocatalytic degradation of chlorinated ethanes was studied in a tubular photoreactor packed with TiO(2) pellets prepared by sol-gel method. The steady-state condition was not obtained, but the deterioration in the photocatalytic activity was observed during the irradiation. Effects of
F Ralph Berberich et al.
Pediatrics, 124(2), e203-e209 (2009-07-15)
The goal was to test a multifaceted distraction method designed to reduce injection-associated pain in school-aged children. A clinical trial evaluated 41 children, 4 to 6 years of age, who were given 3 standard prekindergarten immunizations; 21 were assigned randomly
Xi-Guang Wei et al.
The Journal of organic chemistry, 75(12), 4212-4217 (2010-05-21)
The enhanced reactivity exhibited by six pseudo-alpha-bases, RC[triple bond]CZ(-) (R = H and Cl; Z = O, S, and Se) in gas-phase E2 reactions with ethyl chloride was examined at the G2(+) level. It is found that anomalous reactivity is
Amir Gal-Oz et al.
Dermatology (Basel, Switzerland), 221(4), 373-377 (2010-11-26)
Ethyl chloride (EC) is usually used as a topical anesthetic spray agent. However, its antipruritic effects have never been studied, to the best of our knowledge. A double-blind placebo-controlled prospective study. Overall, 51 healthy volunteers underwent a histamine skin prick

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service