Skip to Content
Merck
All Photos(1)

Key Documents

40012

Supelco

1,1-Dichloroethane solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4Cl2
CAS Number:
Molecular Weight:
98.96
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

ClC(Cl)C

InChI

1S/C2H4Cl2/c1-2(3)4/h2H,1H3

InChI key

SCYULBFZEHDVBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miklós Lakatos
Journal of pharmaceutical and biomedical analysis, 47(4-5), 954-957 (2008-04-26)
The purge-and-trap (P&T) gas extraction method combined with gas chromatography was studied for its suitability for quantitative residual solvents determination in a water-soluble active pharmaceutical ingredient (API). Some analytical method performance characteristics were investigated, namely, the repeatability, the accuracy and
Lewis Semprini et al.
Journal of contaminant hydrology, 103(3-4), 157-167 (2008-11-22)
A field study was performed to evaluate the potential for in-situ aerobic cometabolism of 1,1,1-trichloroethane (1,1,1-TCA) through bioaugmentation with a butane enrichment culture containing predominantly two Rhodococcus sp. strains named 179BP and 183BP that could cometabolize 1,1,1-TCA and 1,1-dicholoroethene (1,1-DCE).
Simon M Pimblott et al.
The journal of physical chemistry. A, 109(45), 10294-10301 (2006-07-13)
The yields of chloride ion and molecular hydrogen were determined in the gamma, the fast electron, and the 5 MeV helium ion radiolysis of deaerated and aerated aqueous solutions of 1,1- and 1,2-dichloroethane. In deaerated solutions irradiated with gamma-rays or
Nancy Vanstone et al.
Environmental science & technology, 42(1), 126-132 (2008-03-21)
Degradation of 1,1- and 1,2-dichloroethane (1,1-DCA, 1,2-DCA) and carbon tetrachloride (CCl4) on Zn0 was investigated using compound specific isotope analysis (CSIA) to measure isotopic fractionation factors for chloroalkane degradation by hydrogenolysis, by alpha-elimination, and by beta-elimination. Significant differences in enrichment
Babu P Patlolla et al.
International journal of environmental research and public health, 2(1), 101-106 (2006-05-19)
The major concern for the halogenated compounds is their widespread distribution, in addition to occupational exposures. Several chlorinated alkanes and alkenes were found to induce toxic effects. In this study, we investigated the genotoxic potential of 1,1-dichloroethane in the bone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service