Skip to Content
Merck
All Photos(1)

Key Documents

36824

Supelco

Haloxyfop-P

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(R)-2-[4-(3-Chloro-5-(trifluoromethyl)-2-pyridyloxy)phenoxy]propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€69.10

€69.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€69.10

About This Item

Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Beilstein:
8571970
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€69.10


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

C[C@@H](Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1)C(O)=O

InChI

1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/t8-/m1/s1

InChI key

GOCUAJYOYBLQRH-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Haloxyfop-P is the stereospecific R-isomer and a metabolite of haloxyfop, that belongs to the group of aryloxyphenoxypropionic acid herbicides. It is used for post-emergence control of annual and perennial grass weeds in sugar beets, fodder beets, oilseed rape, potatoes, leafy vegetables, onions, flax, sunflowers, soybeans, vines, strawberries, and other crops. Haloxyfop-P is absorbed by leaves and roots and hinders fatty acid biosynthesis by inhibiting the Acetyl CoA carboxylase (ACCase) enzyme.

According to Regulation (EC) 1107/2009, repealing Council Directive 91/414/EEC, haloxyfop-P is approved for use as an herbicide in European Union (EU) for post-emergence foliar applications on carrots, fodder legumes (beans, peas dry), rape seeds, soya beans, and sugar beets. The maximum residue level (MRL) for Haloxyfop (Sum of haloxyfop, its esters, salts, and conjugates expressed as haloxyfop (sum of the R- and S- isomers at any ratio)) is set from 0.01 mg/kg to 0.4 mg/kg for products of plant and animal origin according to Regulation (EU) 2017/1016.

Haloxyfop-P has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The haloxyfop-P (also known as haloxyfop-R) CRM may also find following uses:
  • Evaluate the efficacy of the herbicide haloxyfop-R isolated and associated with 2,4-D (DMA 806 BR) in the control of volunteer RR maize at different stages
  • Development of an analytical method to determine residues of haloxyfop-R and its methyl ester in soils and soybeans using gas-liquid chromatography (GLC) with an electron capture detector (ECD)
  • Evaluate the effects of herbicides associated with cover crops (Pennisetum glaucum, Crotalaria spectabilis, and Urochloa ruziziensis) in soybean desiccation management for volunteer RR corn control

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service