Skip to Content
Merck
All Photos(1)

Key Documents

33097-U

Supelco

trimethylanilinium hydroxide (TMAH)

0.2 M in methanol

Synonym(s):

Trimethylphenylammonium hydroxide solution, Phenyltrimethylammonium hydroxide, TMAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(OH)C6H5
CAS Number:
Molecular Weight:
153.22
Beilstein:
3917033
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for GC derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 1 × 10 mL (33097-U)

concentration

0.2 M in methanol

SMILES string

[OH-].C[N+](C)(C)c1ccccc1

InChI

1S/C9H14N.H2O/c1-10(2,3)9-7-5-4-6-8-9;/h4-8H,1-3H3;1H2/q+1;/p-1

InChI key

HADKRTWCOYPCPH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylanilinium hydroxide (TMAH) is an alkylating agent, which is effectively used esterification of carboxylic acids, etherfication of alcohols and N-alkylation of amino groups.

Application

It is used to derivatize the serum sample for determination of Topiramate in Epileptic patients using GC-MS analysis.

Other Notes

Reagent for carboxylic acid methyl ester, methyl esters.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.W. Robb and J.J. Westbrook III
Analytical Chemistry, 35, 1644-1644 (1963)
A capillary GC method using nitrogen phosphorus detection for determination of topiramate in patients with epilepsy.
Malakova, J., et al.
Chromatographia, 66, 363-367 (2007)
Vladimir Zaikin, John M. Halket
A Handbook of Derivatives for Mass Spectrometry, 218-219 (2009)
A Ranz et al.
Journal of biochemical and biophysical methods, 69(1-2), 3-14 (2006-03-28)
In the present study, a derivatization method for the determination of acidic herbicides has been investigated. This procedure involves the methylation with the quaternary ammonium salt trimethylanilinium hydroxide (TMAH) directly in the gas chromatographic auto-sampler vial for analysis by gas
Timothy J Jensen et al.
Journal of photochemistry and photobiology. B, Biology, 100(2), 100-111 (2010-06-19)
Five cationic porphyrins bearing one to four -N(CH(3))(3)(+) groups linked to the p-phenyl positions of 5,10,15,20-tetraphenylporphyrin (TPP) were synthesized in order to study the effect of overall charge and its distribution on the cellular uptake, phototoxicity and intracellular localization using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service