Skip to Content
Merck
All Photos(1)

Key Documents

28878-U

Supelco

Super Clean (Base-Plate Design) Kit

carrier gas kit (includes SU861026 + SU861011)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54
Pricing and availability is not currently available.

General description

Super Clean base-plate purifiers are a unique point-of use glass/metal, diffusion proof purification system to purifiy carrier, fuel, and other gases for the GC or GC-MS system. These purifiers remove hydrocarbons, oxygen (color indicated), and moisture (color indicated) to better than 6.0 gas (99.9999%) quality at 2 L/min., independent of the original gas quality.

Legal Information

Super Clean is a trademark of Scientific Glass Technology B.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service