Skip to Content
Merck
All Photos(1)

Key Documents

2875901-U

Supelco

Inlet Liner, On-Column Type, Straight Design (unpacked)

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115719
NACRES:
SB.54

description

PTV liner for Trace 2000

packaging

pkg of 1 ea

L × O.D. × I.D.

120 mm × 2.75 mm × 2.0 mm
120 mm × 2.75 mm × 2.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Thermo (PTV Injector)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service