28159-U
Omega Battery Charger
AC/DC input 120 V / 240 V AC, units charged: 5
Synonym(s):
Five Unit Omega Charger/Adapter for Q-MAX Air Sampling Pumps, 110/220VAC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with Escort Elf Air Sampling Pump
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service