27214-U
Crimp seals with PTFE/silicone septa
silver aluminum seal, size × thickness 11 mm × 1.0 mm, white PTFE/silicone septum, pkg of 100 ea
Synonym(s):
PTFE/Silicone Septum Crimp Seals
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
silver aluminum seal
white PTFE/silicone septum
feature
closure type crimp seal
packaging
pkg of 100 ea
size × thickness
11 mm × 1.0 mm
fitting
thread for 11 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Crimp seals with PTFE/silicone septa may be used to seal the water samples during the analysis of BTEX in BTEX contaminated waters, using static headspace extraction, followed by gas chromatography with photoionization detection (HS-GC-PID).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Engineering and microbiological aspects of BTEX removal in bioreactors under sulfate-reducing conditions.
Chemical Engineering Journal, 260, 503-512 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service