Skip to Content
Merck
All Photos(1)

Documents

24940-U

Supelco

Purge/Trap K

Vocarb® 3000, for use with OI Analytical Eclipse 4660, 4560

Synonym(s):

PURGE TRAP K, O.I. ANALYTICAL 4560

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NB.21

product name

Purge/Trap K Vocarb® 3000, for use with OI Analytical Eclipse 4660, 4560

Agency

suitable for EPA 601

application(s)

environmental

compatibility

for use with OI Analytical Eclipse 4660, 4560

Looking for similar products? Visit Product Comparison Guide

General description

Purge/Traps K Vocarb® 3000 (configured for OI Analytical Eclipse 4660, 4560) are used to trap the analytes efficiently in US EPA Methods 502.2 and 524.

Legal Information

Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Chromatograms

suitable for GCsuitable for GCsuitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service