Key Documents
24417
SGE Syringes for Agilent/HP 7673 and 7683 Autosampler, Removable Needle
volume 10 μL, needle size 26 ga, needle L 42 mm
About This Item
UNSPSC Code:
41122001
material
cone tip stainless steel needle (point style 1)
needle size
26 ga
description
10R-HP-0.47
color (dark orange)
feature
needle type R (removable needle)
syringe type Standard syringe, removable needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 002805
needle L
42 mm
volume
10 μL
Choose from one of the most recent versions:
Certificates of Analysis (COA)