24110UINT
SP®-2380 Intuvo Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm, for use with Agilent Intuvo GC
Synonym(s):
SP®-2380 Intuvo Capillary GC Column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
fused silica column
df
0.20 μm
technique(s)
gas chromatography (GC): suitable
L × I.D.
30 m × 0.25 mm
column type
capillary highly polar
compatibility
for use with Agilent Intuvo GC
Looking for similar products? Visit Product Comparison Guide
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service