Skip to Content
Merck
All Photos(1)

Key Documents

23817

Supelco

Polyimide Sealing Resin

pkg of 5 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

packaging

pkg of 5 g

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

General description

A GlasSeal connector will form a perfect seal between two fused silica columns. To make this connection extremely durable, use a small drop of this resin. Also for use as an excellent high temperature glue. Cures at 200 °C. For use at 350 °C or lower operating temperatures. The bottle contains 5 g of resin, and includes a handy applicator cap.

Application

Polyimide sealing is used to make a permanent and leak-free union between two pieces of silica tubing, stainless steel or glass, press fit unions are most commonly used.[1]

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

187.0 °F - closed cup

Flash Point(C)

86.11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Emiliano Zampetti et al.
Sensors (Basel, Switzerland), 21(1) (2021-01-01)
Benzene, toluene and xylene (BTX) are an important part of the volatile organic compounds (VOCs) to be detected and monitored in the air, due to their toxicity towards human health. One of the most reliable technique used in BTX detection
Causes and prevention of column damage.
Rood D.
The Troubleshooting and Maintenance Guide for Gas Chromatographers, 228-229 (2007)
Jessica Wade et al.
Nature communications, 11(1), 6137-6137 (2020-12-03)
Polymer thin films that emit and absorb circularly polarised light have been demonstrated with the promise of achieving important technological advances; from efficient, high-performance displays, to 3D imaging and all-organic spintronic devices. However, the origin of the large chiroptical effects
Maxwell Wei-Hao Li et al.
Journal of chromatography. A, 1620, 461002-461002 (2020-03-08)
Micro gas chromatography (µGC) is a technique developed for rapid, in situ analysis of volatile organic compounds (VOCs) for environmental protection, industrial monitoring, and toxicology. While reduced µGC size and power requirements allow for increased portability, the low moisture and

Questions

1–3 of 3 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is a GlasSeal connector?

    1 answer
    1. A GlasSeal connector is a small piece of drawn glass tubing that is used to connect fused-silica capillary columns.  When you want the seal to be permanent, you can use the 23817: Polyimide Sealing Resin

      Helpful?

  3. What is the solvent used with the Polyimide Sealing Resin, Product 23817?

    1 answer
    1. The Supelco 23817: Sealing resin is solubilized in N-Methylpyrrolidone (Aldrich 443778).  Once this liquid polymer solidifies on drying or curing with heat, it can only be degraded by hot N-Methylpyrrolidone or hot concentrated sulfuric acid.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service