Skip to Content
Merck
All Photos(1)

Key Documents

23470-U

Supelco

Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, Propyne, and n-Butane

15 ppm each component in nitrogen, cylinder of 48 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

form

gas

packaging

cylinder of 48 L

manufacturer/tradename

Scott Gas T54-48

availability

not available in EU

concentration

15 ppm each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, Propyne, and n-Butane 15 ppm each component in nitrogen, is a multi component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Analyte

Description

    Acetylene
    Butane
    Ethane
    Ethylene
    Methane
    Propane
    Propylene
    Propyne
    See All (8)

regulator

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service