Skip to Content
Merck
All Photos(1)

Key Documents

23402

Supelco

Carbon dioxide (99.8%)

cylinder of 14 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO2
Molecular Weight:
44.01
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Assay

99.8%

form

gas

packaging

cylinder of 14 L

manufacturer/tradename

Air Liquide America Specialty Gases LLC T105-14

availability

not available in EU

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

SMILES string

O=C=O

InChI

1S/CO2/c2-1-3

InChI key

CURLTUGMZLYLDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Specifications:
Contents
: 14 liters
Pressure: 240 psig
Outlet fitting: CGA-160-1/8 inch, NPT F
Weight: 1.5 lb.
Dimensions
: 3 x 11 inches
D.O.T. specifications: 4B240

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • Carbon dioxide (99.8%) is a speciality pure gas suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Piercy et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 185(8), 891-903 (2015-08-20)
The present study determined whether EEG and/or EMG recordings could be used to reliably define activity states in the Brazilian black and white tegu lizard (Tupinambis merianae) and then examined the interactive effects of temperature and activity states on strategies
Xiaoqing Lu et al.
Blood, 125(4), 649-657 (2014-11-09)
Human germinal center-associated lymphoma (HGAL) is specifically expressed only in germinal center (GC) B lymphocytes and GC-derived lymphomas. HGAL protein decreases lymphocyte motility by inhibiting the ability of myosin to translocate actin via direct interaction with F-actin and myosin II
L Rienzi et al.
Reproductive biomedicine online, 30(1), 57-66 (2014-12-03)
Recent studies involving a limited number of patients have indicated a correlation between aneuploidy and various morphokinetic parameters during preimplantation development. The results among different groups, however, have been inconsistent in identifying the parameters that are able to predict chromosomal
Erik H J Hulzebos et al.
Medicine and science in sports and exercise, 46(11), 2047-2052 (2014-05-23)
Lung function, nutritional status, and parameters of exercise capacity are known predictors of mortality in patients with cystic fibrosis (CF). The aim of the current study was to use these important parameters to develop a multivariate model to predict mortality
Vagner R R Silva et al.
Nature communications, 5, 4859-4859 (2014-09-26)
Sphingosine 1-phosphate receptor 1 (S1PR1) is a G-protein-coupled receptor for sphingosine-1-phosphate (S1P) that has a role in many physiological and pathophysiological processes. Here we show that the S1P/S1PR1 signalling pathway in hypothalamic neurons regulates energy homeostasis in rodents. We demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service