Skip to Content
Merck
All Photos(1)

Documents

22997-U

Supelco

Union

for connecting 1/16 in., bore size 0.75 mm

Synonym(s):

Valco® Unions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

stainless steel

description

internal

packaging

pkg of 1 ea

manufacturer/tradename

VICI Valco® ZU1

bore size

0.75 mm

fitting

for 1/16 in. O.D. tubing

compatibility

for connecting 1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

Stainless steel, unless noted otherwise. Nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service