Skip to Content
Merck
All Photos(1)

Documents

22686

Swagelok® Union Cross

Swagelok® 400-4, brass, 1/4 in. Swagelok

Synonym(s):

Swagelok® 400-4,BRASS UNION CROSS 1/4" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

brass

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 400-4

fitting

1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Union cross generally divides the eluent flow into three directions which is coming from the pump. In gas system, pure gases from mass flow controllers are converged at the union cross and in turn flow into different size tubing to generate turbulence which produce homogenized gas.
Nuts and ferrules included.

Application

Swagelok® union cross was used in sensor test module along with electrical breakout box to pressurized high temperature reactors such as high performance reactor (HPR) microreactor.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen monitoring for power plant applications using SiC sensors.
Loloee, Reza, et al.
Sensors and Actuators B, Chemical, 129, 200-210 (2008)
Synthesis and Characterization of Nano-Composite Alumina-Titania Ceramic Membrane for Gas Separation.
Ahmad, A. L., M. R. Othman, and N. F. Idrus.
Journal of the American Ceramic Society. American Ceramic Society, 89, 3187-3793 (2006)
Katie C Coate et al.
Cell metabolism, 32(6), 1028-1040 (2020-11-19)
Isolated reports of new-onset diabetes in individuals with COVID-19 have led to the hypothesis that SARS-CoV-2 is directly cytotoxic to pancreatic islet β cells. This would require binding and entry of SARS-CoV-2 into β cells via co-expression of its canonical
Chuan-Ming Xie et al.
Autophagy, 10(9), 1495-1508 (2014-06-24)
EEF2K (eukaryotic elongation factor-2 kinase), also known as Ca (2+)/calmodulin-dependent protein kinase III, functions in downregulating peptide chain elongation through inactivation of EEF2 (eukaryotic translation elongation factor 2). Currently, there is a limited amount of information on the promotion of
J.F.K. Huber
Instrumentation for High Performance Liquid Chromatography, 13, 52-52 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service