Skip to Content
Merck
All Photos(1)

Key Documents

21568-U

Supelco

Supelite DAX-8

jar of 5000 g

Synonym(s):

XAD-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.52

form

solid

packaging

jar of 5000 g

technique(s)

HPLC: suitable
LPLC: suitable
ion chromatography: suitable

surface area

140 m2/g

matrix

acrylic ester

matrix active group

polymer

particle size

40-60 mesh

pore size

~0.79 mL/g pore volume
225 Å mean pore size

density

1.09 g/mL at 25 °C (true wet)(lit.)
1.23 g/mL at 25 °C (skeletal)(lit.)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Supelite DAX-8 is a nonfunctionalised resin with strong hydrophobic organic matter endorsed to humic and fulvic acids. It is also referred as polymethylmethacrylate resin.

Application

Adsorbent resin with moderate polarity for compounds up to 150,000 MW. Most commonly used for adsorption of fulvic and humic acids, treatment of paper pulp mill waste, and removal of alcohols, surfactants, and colorants from samples.
Supelite DAX-8 was used as an absorbent in:
  • HPLC-SEC fractionation system
  • column chromatography
  • extraction procedure and these extracts were analysed using Cary Eclipse fluorescence spectrophotometer

Preparation Note

DAX-8 is a salt imbibed resin, which requires some preparation prior to use. In the protocol below, you may use methanol in place of ethanol.
  • First, rinse the adsorbent resin with 2-3 bed volumes (BV) of distilled water.
  • Soak overnight in 2 BV of 95% ethanol.
  • After soaking, the resin is introduced into the column and rinsed with an additional 2 BV of 95% ethanol.
  • Rinse with 2 BV of distilled water to displace the ethanol, followed by 1 BV of 4% (w/v) sodium hydroxide, 2 BV of distilled water, 1 BV 4% (v/v) hydrochloric acid, and finally by distilled water until the pH of the eluent becomes neutral.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The fouling of microfiltration membranes by NOM after coagulation treatment
Carroll, T., et al.
Water Research, 34, 2861-2868 (2000)
Hua Zhang et al.
The Science of the total environment, 407(2), 879-886 (2008-11-15)
Dissolved organic matter (DOM) in leachate samples collected at Laogang Sanitary Landfill in Shanghai, China were fractionated using size exclusion chromatography and were characterized using estrogen receptor-alpha competitor screening assay as toxicity activity index. The estrogen activity of leachate was
Sorption of aquatic humic matter by DAX-8 and XAD-8 resins: comparative study using pyrolysis gas chromatography
Peuravuori, Juhani, Tero Lehtonen, and Kalevi Pihlaja.
Analytica Chimica Acta, 471, 219-226 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service