Skip to Content
Merck
All Photos(1)

Key Documents

18380

Supelco

N-Sulfocarbamoylsaxitoxin solution

20 μg/g in acetic acid, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

GTX5 solution, Gonyautoxin solution, Toxin B1 solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
€296.00

€296.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 ML
€296.00

About This Item

Empirical Formula (Hill Notation):
C10H17N7O7S
CAS Number:
Molecular Weight:
379.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€296.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in acetic acid

format

single component solution

storage temp.

−20°C

SMILES string

[S](=O)(=O)(NC(=O)OC[C@@H]1NC(=N)N2[C@]3(NC(=N)N[C@H]31)C(CC2)(O)O)O

InChI

1S/C10H17N7O7S/c11-6-14-5-4(3-24-8(18)16-25(21,22)23)13-7(12)17-2-1-9(19,20)10(5,17)15-6/h4-5,19-20H,1-3H2,(H2,12,13)(H,16,18)(H3,11,14,15)(H,21,22,23)/t4-,5-,10-/m0/s1

InChI key

JKKCSFJSULZNDN-HGRQIUPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service