Skip to Content
Merck
All Photos(1)

Key Documents

11101AST

Supelco

Astec® CHIROBIOTIC® V Chiral (5 μm) HPLC Columns

L × I.D. 2 cm × 1 mm, HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CHIROBIOTIC® V Chiral HPLC Guard Column, Column, 5 μm particle size, L × I.D. 2 cm × 1 mm

material

stainless steel column

Agency

suitable for USP L88

description

HPLC guard column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 1 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

vancomycin phase

particle size

5 μm

pore size

100 Å

operating pH range

3.5-7.0

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin-xia Chen et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 50(4), 486-491 (2015-08-01)
A chiral LC-MS/MS method for the simultaneous analysis of desvenlafaxine (DVS) enantiomers in human plasma was developed and applied to a pharmacokinetic study on 12 Chinese healthy volunteers. d6-Desvenlafaxine was used as internal standard (IS). Chromatographic separation was performed on
Shu Jin et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 47(1), 105-109 (2012-04-12)
To study the drug-drug interaction of morinidazole and warfarin and its application, a sensitive and rapid liquid chromatography-tandem mass spectrometric (LC-MS/MS) method was developed for the determination of R-warfarin/S-warfarin in human plasma. In a random, two-period crossover study, 12 healthy
Chenghao Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1011, 45-52 (2016-01-12)
A chiral UFLC-MS/MS method was established and validated for quantifying d-threo-methylphenidate (d-threo-MPH), l-threo-methylphenidate (l-threo-MPH), d-threo-ethylphenidate (d-threo-EPH), l-threo-ethylphenidate (l-threo-EPH) and d,l-threo-ritalinic acid (d,l-threo-RA) in rat plasma over the linearity range of 1-500ng/mL. Chiral separation was performed on an Astec Chirobiotic V2
Osama Y Alshogran et al.
Journal of chromatographic science, 54(4), 554-560 (2015-12-15)
A simple and rapid liquid chromatographic-tandem mass spectrometric method has been developed and validated for the enantiospecific determination of R- and S-warfarin in human urine. Warfarin enantiomers were extracted from urine using methyl tert-butyl ether. Chromatographic separation of warfarin enantiomers
Thomas Kraemer et al.
Journal of analytical toxicology, 27(2), 68-73 (2003-04-03)
Prenylamine (R,S-N-(3,3-diphenylpropyl-methyl-2-phenethylamine), a World Health Organization class V calcium antagonist, is known to be metabolized to amphetamine. In this study, amphetamine concentrations after a single-dose administration of prenylamine were determined to check if they reached values that could be of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service