Skip to Content
Merck
All Photos(12)

Key Documents

ZRB69

Sigma-Aldrich

Anti-Nestin Antibody, clone 8B8, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

NES

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8B8, monoclonal
recombinant monoclonal

description

8B8 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 177.44 kDa
observed mol wt ~_220 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry: suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NES(10763)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 8B8 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Nestin. It targets an epitope with in 150 amino acids from the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 150 amino acids from the C-terminal region of human Nestin.

Application

Anti-Nestin, clone 8B8, ZooMAb is a recombinant Rabbit Monoclonal antibody that detects Nestin and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Nestin in SH-SY5Y cells.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Nestin in lysate from Human H9 Stem Cells.

Target description

Nestin (UniProt: P48681) is encoded by the NES (also known as Nbla00170) gene (Gene ID: 10763) in human. Nestin is a class VI intermediate filament protein that is required for survival, renewal, and mitogen-stimulated proliferation of neural progenitor cells. Nestin expression is up-regulated in most mitotic cells and is downregulated upon differentiation. Upon terminal neural differentiation Nestin levels are shown to be down-regulated and replaced by neurofilaments. Nestin is also shown to be expressed in kidney in a developmentally regulated pattern. Its expression is also associated with tubulointerstitial injury and can serve as a marker for peritubular endothelial cell injury and tubulointerstitial fibrosis. Nestin is reported to be constitutively phosphorylated and its phosphorylation state increases during mitosis when the cytoplasmic intermediate filament network is reorganized. Nestin has a short N-terminus and an unusually long C-terminus region that results in complex binding to an array of structural proteins. The short N-terminus leads to preferential intermediate filaments formation with heterodimers and heterotetramers and the long C-terminal portion functions as links or bridges between intermediate filaments and microtubules. Nestin can exist in homodimeric or homotetrameric form, however in mixtures with other intermediate filament proteins such as vimentin and alpha-internexin, it can form heterodimers, which can assemble to form intermediate filaments. Nestin is shown to promote the disassembly of phosphorylated vimentin intermediate filaments during mitosis and play a role in the trafficking and distribution of intermediate filament proteins to daughter cells during progenitor cell division. (Ref.: Xie, L., et al. (2015). Stem Cells Int. 2015:762098; Tomioka, M., et al. (2010). 15(5); 568-574).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service