Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2680

Sigma-Aldrich

Anti-PAFAH/PLA2G7 Antibody, clone 1E9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

1-alkyl-2-acetylglycerophosphocholine esterase, 2-acetyl-1-alkylglycerophosphocholine esterase, EC:3.1.1.47, Group-VIIA phospholipase A2, LDL-PLA(2), LDL-associated phospholipase A2, PAF 2-acylhydrolase, PAF acetylhydrolase, Platelet-activating factor acetylhydrolase, gVIIA-PLA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 50.08 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

rat, human, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PLA2G7(7941)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E9 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Platelet-activating factor acetylhydrolase (PAFAH). it targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human Platelet-activating factor acetylhydrolase (PAFAH).

Application

Quality Control Testing

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PAFAH/PLA2G7 in RAW264.7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PAFAH/PLA2G7 in lysates from HL-60 cells, Human spleen tissue and Rat spleen tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PAFAH/PLA2G7 in Human spleen tissue sections.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected PAFAH/PLA2G7 in Human spleen tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound PAFAH/PLA2G7 peptide with a KD of 1.1 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Platelet-activating factor acetylhydrolase (UniProt: Q13093; also known as EC:3.1.1.47, PAF acetylhydrolase, 1-alkyl-2-acetylglycerophosphocholine esterase, 2-acetyl-1-alkylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), PAF 2-acylhydrolase) is encoded by the PLA2G7 (also known as PAFAH) gene (Gene ID: 7941) in human. PAFAH/PLA2G7 is a lipoprotein-associated calcium-independent A2 involved in phospholipid catabolism during inflammatory and oxidative stress response. It is a secreted protein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce the mature protein. In plasma, it is reported to associates with both LDL and HDL particles. At the lipid-aqueous interface it is shown to hydrolyze the ester bond of fatty acyl group attached at sn-2 position of phospholipids. It specifically targets phospholipids with a short-chain fatty acyl group at sn-2 position, however, it can hydrolyze phospholipids with long fatty acyl chains, only if they carry oxidized functional groups. PAFAH/PLA2G7 also hydrolyzes and inactivates platelet-activating factor (PAF), a potent pro-inflammatory signaling lipid that acts through PTAFR on various innate immune cells. It also hydrolyzes oxidatively truncated phospholipids carrying an aldehyde group at the omega position, preventing their accumulation in low-density lipoprotein (LDL) particles and uncontrolled pro-inflammatory effects. Mutations in PLA2G7 gene have been linked to Platelet-activating factor acetylhydrolase deficiency (PAFAD) that leads to impaired response to inflammatory agents and leads to inflammatory gastrointestinal disorders, asthma, and atopy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Stafforini, DM., et al. (1999). J. Biol. Chem. 274(11); 7018-7024; Tjoelker, LW., et al. (1995). Nature. 374(6522); 549-553).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service