Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2660

Sigma-Aldrich

Anti-PTPRZ1 Antibody, clone 1J4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

PTPRZ, Protein-tyrosine phosphatase receptor type Z polypeptide 1, Protein-tyrosine phosphatase receptor type Z polypeptide 2, R-PTP-zeta, R-PTP-zeta-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 255 kDa
observed mol wt ~270 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PTPRZ1(5803)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J4 is a ZooMAb® rabbit monoclonal that specifically detects PTPRZ1. It targets an epitope within 24 amino acids from the N-terminal half of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human PTPRZ1.

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Receptor-type tyrosine-protein phosphatase zeta (PTPRZ1) in A549 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PTPRZ1 in HUVEC and NIH/3T3 cell lysates and a 1:10,000 dilution detected PTPRZ1 in human brain tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PTPRZ1 in human brain tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PTPRZ1 in A549 cells.

Affinity Binding Assay; The binding affinity of this antibody to PTPRZ1-Phosphocan peptide was at least one billion-fold (1,000,000,000X) higher than that obtained with the non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Receptor-type tyrosine-protein phosphatase zeta (UniProt: P23471; also known as EC:3.1.3.48, R-PTP-zeta, Protein-tyrosine phosphatase receptor type Z polypeptide 1, Protein-tyrosine phosphatase receptor type Z polypeptide 2, R-PTP-zeta-2) is encoded by the PTPRZ1 (also known as HTPZP2, PTPRZ, PTPRZ2, PTPZ) gene (Gene ID: 5803) in human. PTPRZ1is a single-pass transmembrane glycoprotein that belongs to the receptor-type PTP (RPTP) protein subfamily. Its expression is observed mainly in the central nervous system, where it is localized in the Purkinje cell layer of the cerebellum, the dentate gyrus, and the subependymal layer of the anterior horn of the lateral ventricle. It is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to produce the mature form that contains a large extracellular domain (aa 25-1636), a transmembrane domain (aa 1637-1662) and a cytoplasmic domain (aa 1663-2315). A secreted Isoform is also reported that is generated by shedding of the extracellular domain. PTPRZ1 negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord and is required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. PTPRZ1 is involved in regulating cell adhesion, migration, establishment of contextual memory and axon guidance through the dephosphorylation of tyrosine residues in various signaling pathways. It may also play a role in protecting oligondendrocytes against apoptosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service