Skip to Content
Merck
All Photos(4)

Key Documents

ZRB2520

Sigma-Aldrich

Anti-PKD/PKCμ Antibody, clone 1C11 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.11.13, Protein kinase C mu type, Protein kinase D, Serine/threonine-protein kinase D1, nPKC-D1, nPKC-mu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 101.70 kDa
observed mol wt ~115 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PRKD1(5587)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C11is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects PKD/PKC . It targets an epitope within 18 amino acids from the C-terminal region of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human PKD/PKC .

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from LNCaP cells.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected PKD/PKC in lysate from LNCaP cells.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected PKD/PKC in lysate from HuH-7 cells.

Affinity Binding Assay: A representative lot of this antibody bound with PKD/PKC peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PKD/PKC in LNCaP cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PKD/PKC in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Serine/threonine-protein kinase D1 (UniProt: Q15139; also known as Protein kinase C mu type, EC:2.7.11.13, Protein kinase D, nPKC-D1, nPKC-mu) is encoded by the PRKD1 (also known as PKD, PKD1, PRKCM ) gene (Gene ID: 5587) in human. The PRKD1 gene promoter has a CpG island covering 1.2 kb and It includes, the transcription start site and the exon 1. The Serine/threonine-protein kinase D1 protein structure features a conserved catalytic Protein Kinase domain at the C-terminus and, an N-terminal regulatory domain that includes a diacylglycerol (DAG)/phorbol ester-binding C1 domain and a pleckstrin homology (PH) domain. Typically, the protein is found in the cytoplasm; however, upon activation, it can translocate to the plasma membrane, golgi appartaus or mitochondria. This translocation is crucial for targeting specific substrates and implies a spatial regulation of its kinase activity. PRKD1 is ubiquitously expressed in various human tissues, with significant expression in the heart, skeletal muscle, and brain. It is also expressed in cells of the normal mammary gland. The protein is involved in many cellular processes, including Golgi body membrane integrity and transport, cell migration and differentiation, MAPK8/JNK1 and Ras pathway signaling, MAPK1/3 (ERK1/2) pathway signaling, vesicle transport, cell survival, and regulation of cell shape and adhesion. In mammary glands, it preserves the epithelial phenotype by inhibiting epithelial-to-mesenchymal transition. Overexpression and dysregulation of PRKD1 are observed in several cancers, including prostate, gastric, and pancreatic cancers, suggesting a role in tumor progression and metastasis. Moreover, PRKD1 has been linked to cardiovascular diseases and neurodegenerative disorders, where its signaling pathways impact cell survival and function. There is also emerging evidence suggesting PRKD1′s role in immune system modulation, potentially impacting autoimmune diseases and inflammation. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref; Zhang W., et al., J Biol Chem 2005; 280(19): 19036-19044; Hurd C., et al., Oncogene 2002; 21(14): 2154-60.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service