Skip to Content
Merck
All Photos(10)

Key Documents

ZRB2342

Sigma-Aldrich

Anti-Fas/CD95 Antibody, clone 1F17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Apo-1 antigen, Apoptosis-mediating surface antigen FAS, CD95, FASLG receptor, Tumor necrosis factor receptor superfamily member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F17, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... FAS(355)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F17 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Fas/CD95. It targets an epitope within 18 amino acids from the cytoplasmic domain in the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Fas/CD95.

Application

Quality Control Testing

Evaluated by Western Blotting in HT-1080 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Fas/CD95 in HT-1080 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Fas/CD95 in Jurkat cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Fas/CD95 in Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound Fas-CD95 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected Fas/CD95 in one million Raji and Jurkat cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Fas/CD95 in Human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tumor necrosis factor receptor superfamily member 6 (UniProt: P25445; also known as Apo-1 antigen, Apoptosis-mediating surface antigen FAS, FASLG receptor, CD95) is encoded by the FAS (also known as APT1, FAS1, TNFRSF6) gene (Gene ID: 355) in human. The interaction of Fas with its ligand FasL (FasL/CD95L) regulates numerous physiological and pathological processes that are mediated through extrinsic apoptotic cell death. It is critical for the maintenance of immune tolerance. Seven isoforms of Fas have been described that are produced by alternative splicing. Isoform 1 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 26-173), a transmembrane domain (aa 174-190), and a cytoplasmic domain (aa 191-335). Isoforms 2 to 6 are secreted forms and isoform 7 is a dominant negative isoform that is resistant to Fas-mediated apoptosis. Isoforms 1 and 6 are expressed at equal levels in resting peripheral blood mononuclear cells; however, after activation there is an increase in isoform 1 and decrease in the levels of isoform 6. The binding of FasL to Fas drives Fas clustering and binding to FADD. Death domain of Fas that is localized in amino acids 230-314 is involved in its binding to FADD. FADD then recruits caspase-8 to form the death-inducing signaling complex (DISC) that is activated by specific post-translational modifications of the death receptor (DR), such as palmitoylation and O-linked glycosylation. The DISC mediates autocatalytic processing and activation of caspase-8, which propagate the death signal through proteolysis of effector other caspases such as caspases-3, 6, and 7. Mutations in FAS gene have been linked to autoimmune lymphoproliferative syndrome 1A that manifests in early childhood and results in the accumulation of autoreactive lymphocytes, which is characterized by non-malignant lymphadenopathy with hepatosplenomegaly, autoimmune hemolytic anemia, thrombocytopenia, and neutropenia. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yamada, A., et al. (2017). Front. Immunol. 8; 403; Scott, FL., et al. (2009). Nature. 457(7232); 1019-1022).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service