ZRB1881
Anti-MCT1/SLC16A1 Antibody, clone 2B21 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Monocarboxylate transporter 1;Solute carrier family 16 member 1
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2B21, recombinant monoclonal
description
2B21 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 53.94 kDa
observed mol wt ~54 kDa
purified by
using Protein A
species reactivity
rat, human
species reactivity (predicted by homology)
monkey
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
Internal
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... SLC16A1(6566)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in L6 cell lysate.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected MCT1/SLC16A1 in L6 cell lysate.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MCT1/SLC16A1 in lysates from Rat skeletal muscle tissue and U-251 MG cells.
Flow Cytometry Analysis: 1 μg from a representative lot detected MCT1/SLC16A1 in one million U-251 MG cells.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MCT1/SLC16A1 in U-251 MG cells.
Affinity Binding Assay: A representative lot of this antibody bound MCT1/SLC16A1 peptide with a KD of 1.7 x 10-8 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western Blotting in L6 cell lysate.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected MCT1/SLC16A1 in L6 cell lysate.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service