Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1733

Sigma-Aldrich

Anti-IDH1 Antibody, clone 1M9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Isocitrate dehydrogenase [NADP] cytoplasmic;EC:1.1.1.42;IDH;Cytosolic NADP-isocitrate dehydrogenase;IDP;NADP(+)-specific ICDH;Oxalosuccinate decarboxylase

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1M9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 46.66 kDa
observed mol wt ~53 kDa

purified by

using Protein A

species reactivity

mouse, rat

species reactivity (predicted by homology)

human, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IDH1(3417)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1M9 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Isocitrate dehydrogenase [NADP] cytoplasmic (IDH1). It targets an epitope within 14 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal half of human Isocitrate dehydrogenase [NADP] cytoplasmic (IDH1).

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse Kidney tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected IDH1 in Mouse Kidney tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IDH1 in C2C12 and NIH3T3 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IDH1 in NIH 3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound IDH1 with a KD of 5.9 x 10-10 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IDH1 in mouse testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Isocitrate dehydrogenase [NADP] cytoplasmic (UniProt: EC:1.1.1.42, IDH, Cytosolic NADP-isocitrate dehydrogenase, IDP, NADP(+)-specific ICDH, Oxalosuccinate decarboxylase) is encoded by the IDH1 (also known as PICD) gene (Gene ID: 3417) in human. A total of five isocitrate dehydrogenases have been described: three of these are NAD(+)-dependent that localize to the mitochondrial matrix, and two are NADP(+)-dependent enzymes. One of the NADP(+) enzyme is mitochondrial and the other is predominantly localized in the cytoplasm. IDH1 is a homodimeric enzyme that is localized in cytoplasm and peroxisomes and catalyzes the oxidative decarboxylation of isocitrate to 2-oxoglutarate. It has two NADP binding sites (aa 75-77 and 310-315) and plays a significant role in cytoplasmic NADPH production. Acetylation at Lys-374 is reported to significantly reduce its catalytic activity. Mutations in IDH1 gene, particularly those affecting Arg 132, have been linked to development of high-grade gliomas. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Reitman, ZJ., et al. (2011). Proc. Natl. Acad. Sci. USA. 108(8); 3270-3275; Dang, L., et al. (2009). Nature. 462(7274); 739-744).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service