Skip to Content
Merck
All Photos(9)

Documents

ZRB1667

Sigma-Aldrich

Anti-p-DRP1-Ser637 Antibody, clone 5G23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5G23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 81.88 kDa
observed mol wt ~75 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

canine, rat, feline, bovine, mouse, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer637)

Gene Information

human ... DNM1L(10059)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5G23 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Dynamin-related protein 1 (DRP1) phosphorylated on serine 637.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 637 from the C-terminal region of human Dynamin-related protein 1 (DRP1).

Application

Quality Control Testing

Evaluated by Western Blotting in a construct containing an N-terminal MBP tagged and C-terminal His tagged recombinant phosphorylated DRP1 (Ser637) protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected a construct containing an N-terminal MBP tagged and C-terminal His tagged recombinant phosphorylated DRP1 (Ser637) protein, but did not react with non-phosphorylated DRP1 construct. (DRP1 construct: Courtesy of Dr. Jesse Rinehart, Yale University, School of Medicine)..

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected p-DRP1-Ser637 in lysate from NIH3T3 cells treated with Forskolin (50 mM)..

Peptide Inhibition Assay: Target band detection in a construct containing an N-terminal MBP tagged and C-terminal His tagged recombinant phosphorylated DRP1 (Ser637) protein fragment was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but non the corresponding non-phosphopeptide.

Affinity Binding Assay: A representative lot of this antibody bound p-DRP1-Ser637 with a KD of 1.3 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Dynamin-1-like protein (UniProt: O00429; also known as EC:3.6.5.5, Dnm1p/Vps1p-like protein, DVLP, Dynamin family member proline-rich carboxyl-terminal domain less, Dymple, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, HdynIV, Dynamin-related protein 1) is encoded by the DNM1L (also known as DLP1, DRP1) gene (Gene ID: 10059) in human. DRP1 is a ubiquitously expressed, homotetrameric protein that plays an important role in mitochondrial fission during mitosis. DRP1 mediates membrane fission through oligomerization into membrane-associated tubular structures that wrap around the scission site to constrict and sever the mitochondrial membrane through a GTP hydrolysis-dependent mechanism. Although DRP1 is essential, but it is not sufficient for mitochondrial fission and requires the recruitment of certain receptors that recruit the enzyme to the outer mitochondrial membrane. In mammals these receptors include MFF, MID49, and MID51. DRP1 phosphorylation/dephosphorylation at two sites near the GED domain (aa 644-735) regulates mitochondrial fission. Its phosphorylation at serine 637 by CaM kinase I and PKA inhibits the GTPase activity that leads to a defect in mitochondrial fission and promotes mitochondrial elongation. Dephosphorylation at this site by protein phosphatase 3 catalytic subunit a (PPP3CA) promotes the fission events. Phosphorylation at serine 616 by Cdk1 and PINK1 activates GTPase and promotes mitochondrial fission. DRP1 is also required for a normal rate of cytochrome c release and caspase activation during apoptosis and this requirement depends upon the cell type and the physiological apoptotic cues. Nine isoforms of DRP1 have been described that are produced by alternative splicing. Isoform 1 with canonical sequence is shown to be brain-specific. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Kalia, R., et al. (2018). Nature. 558(7710). 401-405; Fröhlich, C., et al. (2013). EMBO J. 32(9); 1280-1292; Smirnova, E., et al. (2001). Mol. Biol. Cell. 12(8); 2245-2256).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service