Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1575

Sigma-Aldrich

Anti-TLR4 Antibody, clone 2N20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Toll-like receptor 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 95.68 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TLR4(7099)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2N20 is a ZooMAb® Rabbit recombinant monoclonal antibody that detects Toll-like receptor 4 (TLR4). It targets an epitope within 16 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the extracellular domain within the N-terminal half of Toll-like receptor 4 (TLR4).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human bone marrow tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected TLR4 in human bone marrow tissue sections.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TLR4 in human skin tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound TLR4 with a KD of 1.4 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 1 μg from a representative lot detected TLR4 in one million U2OS cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TLR4 in U2OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Toll-like receptor 4 (UniProt: O00206; also known as EC:3.2.2.6, hToll, CD284, TLR4) is encoded by the TLR4 gene (Gene ID: 7099) in human. Toll-like Receptors (TLRs) are transmembrane proteins that are expressed on various immune cells. They recognize specific molecular patterns associated with pathogenic species and provide an early defense mechanism against foreign organisms. They contain an N-terminal extracellular and a C-terminal cytoplasmic domain. The N-terminal region contains leucine-rich repeats (LRR) that recognize specific pathogen components. Although the ligand recognition mechanisms of the TLRs vary significantly, all TLR dimers exhibit similar overall arrangement. At least 13 different members of TLR family have been identified that detect different pathogen associated molecular patterns (PAMPs). TLR4 is a single-pass type I membrane glycoprotein that is highly expressed in heart, spleen, lung, muscle, and macrophages. It is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 24-631), a transmembrane domain (aa 632-652), and a cytoplasmic domain (aa 653-89). TLR4 cooperates with LY96 and CD14 to mediate the innate immune response to bacterial lipopolysaccharide. It acts via MYD88, TIRAP and TRAF6, leading to NF-kB activation, cytokine secretion and the inflammatory response. In complex with TLR6, it also promotes sterile inflammation in monocytes/macrophages in response to oxidized low-density lipoprotein. It is also shown to be a crucial mediator in oxLDL-induced inflammatory cytokine expression and secretion in smooth muscle cells. TLR4 contains 18 LRR repeats and one TIR domain (aa 672-815) that mediates NAD+ hydrolase activity. D17 (Ref.: Yang, K., et al. (2014). PLoS ONE 9(4); e95935; Akashi, S., et al. (2000). J. Immunol. 164; :3471-3475).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service