Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1481

Sigma-Aldrich

Anti-Phospho-ACC1-(Ser79) Antibody, clone 1E11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Acetyl-CoA carboxylase 1;EC:6.4.1.2;ACC-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 265.19 kDa
observed mol wt ~250 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer79)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E11 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Acetyl-CoA carboxylase 1 (ACC1) phosphorylated on serine 79.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserine 79 from the N-terminal region of rat Acetyl-CoA carboxylase 1 (ACC1).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from NIH3T3 cells overnight starved and treated Oligomycin.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Acetyl-CoA Carboxylase 1 (ACC1) phosphorylated on serine 79 in lysate from NIH3T3 cells overnight starved and treated with 0.5 μM Oligomycin for 30 min.

Tested applications

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected a construct containing GST-tagged ACC1 phosphorylated on serine 79 (Lane 2) but did not react with non-phosphorylated construct (Lane 1). (Phospho-ACC1 construct: Courtesy of Dr. Jesse Rinehart, Yale University, School of Medicine).

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Phospho-ACC1-(Ser79) in human breast cancer tissue sections.

Peptide Inhibition Assay: Target band detection in a lysate from NIH3T3 cells treated with Oligomycin (0.5 mM) was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Acetyl-CoA carboxylase 1 (UniProt: P11497; also known as EC:6.4.1.2, ACC1, ACC-alpha) is encoded by the Acaca (also known as Acac) gene (Gene ID: 60581) in rat. Acetyl-CoA carboxylase 1 (ACC1) is a multi-subunit, biotin-dependent cytosolic enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT). The activity of ACC can be controlled at the transcriptional level as well as by small molecule modulators and covalent modification. Two isoforms of ACC have been reported, alpha and beta that are encoded by two different genes. ACC1 is highly enriched in lipogenic tissues. Its expression is observed in brain, placenta, skeletal muscle, pancreas, and adipose tissue. It is not detected in the liver. Its levels are regulated under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Phosphorylation of ACC1 at serine 79 by AMP-activated protein kinase (AMPK) inhibits its activity. It occurs when glucagon or epinephrine binds to the receptors or when the energy status of the cell is low, which leads to the activation of the AMPK. The presence of fatty acids also inhibits the activities of ACC1. ACC is considered as an important target in the design of new anti-obesity drugs. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service