Skip to Content
Merck
All Photos(18)

Documents

ZRB1417

Sigma-Aldrich

Anti-p-Rb-Ser807/811 Antibody, clone 3J7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Retinoblastoma-associated protein;p105-Rb;p110-RB1;pRb;pp110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3J7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 106.16 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

rat, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer807/pSer811)

Gene Information

human ... RB1(5925)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3J7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Retinoblastoma-associated protein phosphorylated on serine 807 and 811.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids surrounding phosphoserines 807 and 811 from human Retinoblastoma-associated protein (Rb).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cells treated with nocodazole.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-Rb-Ser807/811 in HeLa cells treated with Nocodazole (40 ng/mL; 8 h).

Tested Applications

Flow Cytometry Analysis: 0.1 μg from a representative lot detected phospho-Rb-Ser807/811 in one million serum starved MCF-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Rb-Ser807/811 in C6 cells.

Affinity Binding Assay: A representative lot of this antibody bound phospho-Rb-Ser807-811 peptide with a KD of 9.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-Rb-Ser807/811 in human lung cancer and human ovarian cancer tissue sections.

Peptide Inhibition Assay: Target band detection in a lysate from HeLa cells treated with Nocodazole (40 ng/mL; 8 h) was prevented by pre-blocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Retinoblastoma-associated protein (UniProt: P06400; also known as p105-Rb, p110-RB1, pRb, Rb, pp110) is encoded by the RB1 gene (Gene ID: 5925) in human. Retinoblastoma (Rb) protein act as a negative regulators of cell cycle and controls the passage of cells from G1 to S phase. It is a key regulator of entry into cell division that acts as a tumor suppressor. It acts as a transcription repressor of E2F1 target genes. In its underphosphorylated, active state it interacts with E2F1 and represses its transcription activity, leading to cell cycle arrest. Its action is regulated by phosphorylation of several serine and threonine residues on the protein. Phosphorylated Rb protein sequesters transcription factors allowing the transition from G1 to S phase. It is phosphorylated by Cdk6 and Cdk4, and subsequently by Cdk2 at serine 567 in G1, thereby releasing E2F1 which is then able to activate cell growth. It is dephosphorylated in the late M phase. Phosphorylation at threonine 821 and 826 promotes interaction between the C-terminal domain and the pocket domain (aa 373-771), and thereby inhibits interactions with heterodimeric E2F/DP transcription factor complexes. Phosphorylation of serine 807 and 811 residues by Cdk3/cyclin-C is shown to be essential for G0-G1 transition. Mutations in Rb1 gene are known to cause childhood retinoblastoma, a malignant tumor that arises from the nuclear layers of the retina. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chinnam, M., and Goodrich, DW. (2011). Curr. Top. Dev. Biol. 94; 129-169; Harbour, JW., et al. (1999). Cell. 98(6); 859-869).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service