Skip to Content
Merck
All Photos(6)

Documents

ZRB1381

Sigma-Aldrich

Anti-VAChT Antibody, clone 1J10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Vesicular acetylcholine transporter;Solute carrier family 18 member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 56.9 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SLC18A3(6572)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J10 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detect Vesicular acetylcholine transporter (VAChT). It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Vesicular acetylcholine transporter (VAChT).

Application

Quality Control Testing

Evaluated by Western Blotting with GST-tagged recombinant fragment of Human VAChT.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected GST-tagged recombinant fragment of Human VAChT.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound VAChT peptide with a KD of 4.8 x 10-8 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: Various dilutions from a representative lot detected GST-tagged recombinant VAChT.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Vesicular acetylcholine transporter (UniProt: Q16572; also known as VAChT, Solute carrier family 18 member 3) is encoded by the SLC18A3 (also known as VACHT) gene (Gene ID: 6572) in human. Acetylcholine (ACh) is synthesized in nerve terminals from choline and acetyl coenzyme A by the action of choline acetyltransferase and then it is transported into synaptic vesicles, where it is stored until release. VAChT is a multi-pass membrane glycoprotein with seven cytoplasmic domains, twelve transmembrane domains, and six lumenal vesicle domains. It is expressed in peripheral and central cholinergic nervous systems and is involved in acetylcholine transport into synaptic vesicles. VAChT serves as an antiporter that couples ACh entry into the vesicle with proton efflux that allows for the accumulation of ACh at high concentrations. Overexpression of VAChT is shown to increase ACh at the synaptic cleft and accelerate aging of neuromuscular junction and motor deficits in SOD1 (G93A) mice. Mutations in SLC18A3 gene are known to cause congenital Myasthenic syndrome that is characterized by failure of neuromuscular transmission, including pre-synaptic, synaptic, and post-synaptic disorders that leads to easy fatigability and muscle weakness. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sugita, S., et al (2016). Skeletal Muscle. 6; 31; Arvidsson, U., et al. (1997). J. Comp. Neurol. 378(4); 454-467; Erickson, JD., et al. (1994). J. Biol. Chem. 269(35); 1929-1932).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service