Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1299

Sigma-Aldrich

Anti-KCNQ2 Antibody, clone 2G18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 95.85 kDa
observed mol wt ~150 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2G18 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Potassium voltage-gated channel subfamily KQT member 2 (KCNQ2). It targets an epitope within 20 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal, cytoplasmic domain of human Potassium voltage-gated channel subfamily KQT member 2 (KCNQ2).

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse brain microsomal preparation.

Western Blotting Analysis: 1:10,000 of this antibody detected KCNQ2 in Mouse brain microsomal preparation.

Tested applications


Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected KCNQ2 in human brain tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound KCNQ2 with a KD of 1.0 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected KCNQ2 in E18 rat cortical cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-KCNQ2, clone 2G18 ZooMAb®, Cat. No. ZRB1299, is recombinant Rabbit monoclonal antibody that detects KCNQ2 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Potassium voltage-gated channel subfamily KQT member 2 (UniProt: O43526; also known as KQT-like 2, Neuroblastoma-specific potassium channel subunit alpha KvLQT2, Voltage-gated potassium channel subunit Kv7.2, KCNQ2) is encoded by the KCNQ2 gene (Gene ID: 3785) in human. KCNQ2 is a multi-pass membrane protein with four cytoplasmic domains, six transmembrane domains, and four extracellular domains. It is highly expressed in areas containing neuronal cell bodies and low expression is observed in spinal cord and corpus callosum. KCNQ2 associates with KNNQ3 to form a potassium channel with essentially identical properties to the channel underlying the native M-current, a slowly activating and deactivating potassium conductance, which plays a role in determining the subthreshold electrical excitability of neurons and the responsiveness to synaptic inputs. It is also important in regulation of neuronal excitability. Mutations in KCNQ2 gene are usually inherited in an autosomal-dominant manner in epileptic syndromes and KCNQ2-associated seizures are observed as early as the first week after birth. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lee, IC., et al. (2020). Sci. Rep. 10; Article 13375).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service