Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1291

Sigma-Aldrich

Anti-AFF2 Antibody, clone 1G13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

AF4/FMR2 family member 2;Fragile X E mental retardation syndrome protein;Fragile X mental retardation 2 protein;FMR2P;Protein FMR-2;Protein Ox19.

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G13, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 144.77 kDa
observed mol wt ~175 kDa

purified by

using Protein A

species reactivity

mouse

species reactivity (predicted by homology)

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G13 is a Rabbit recombinant monoclonal antibody that specifically detects AF4/FMR2 family member 2 (AFF2). It targets an epitope within 160 amino acids from the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 160 amino acids from the C-terminal half of human AF4/FMR2 family member 2 (AFF2).

Application

Quality Control Testing

Evaluated by Western Blotting in Neuro2A cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AFF2 in Neuro2A cell lysate.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected AFF2 in mouse brain tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant AFF2 with a KD of 8.6 x 10-9 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

AF4/FMR2 family member 2 (UniProt: P51816; also known as Fragile X E mental retardation syndrome protein, Fragile X mental retardation 2 protein, FMR2P, Protein FMR-2, Protein Ox19) is encoded by the AFF2 (also known as FMR2, OX19) gene (Gene ID: 2334) in human. In mammals four different homologs of Drosophila Lilli protein have been described and are named as AFF1, AFF2, AFF3, and AFF4. AFF1 and 4 are part of super elongation complex (SEC) and are required to release polymerase II. AFF2 and 3 are key subunits of the SEC-like complexes SEC-L2 and SEC-L3, respectively. They are required for transcription elongation in some genes. AFF2 is RNA-binding protein that is involved in alternative splicing regulation through an interaction with G-quartet RNA structure. It associates with mRNA via one of the three RNA-binding domains. AFF2 is mainly expressed in the brain and is most abundant in hippocampus and amygdala. Some expression is also observed in placenta and lung. It localizes with splicing factor SC35 in nuclear speckles where splicing factors are concentrated, assembled, and modified. It is reported to regulates the transcription and toxicity of expanded GGGGCC (G4C2) repeat-containing C9ORF72 allele in Amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). CRISPR-Cas9-mediated loss of AFF2 is shown to reduce levels of C9ORF72 RNA, RNA foci, and DPR proteins in neurons derived from iPSCs of patients with C9ORF72-ALS/FTD. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yuva-Aydemir, Y., et al. (2019). Nat. Commun. 10; 10, Article 5466; Sidorov, MS., et al., (2013). Mol. Brain. 6; Article 15; Melko, M., and Bardoni, B. (2010). Biochemie. 92; 919-926).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service