Skip to Content
Merck
All Photos(21)

Key Documents

ZRB1186

Sigma-Aldrich

Anti-ZNF638 Antibody, clone 4C19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CTCL-associated antigen se33-1, Cutaneous T-cell lymphoma-associated antigen se33-1, Nuclear protein 220, Zinc finger matrin-like protein, Zinc finger protein 638

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Estimated to ship on13 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 220.63 kDa
observed mol wt ~220 kDa

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 4C19 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Zinc Finger Protein 638 (ZNF638). It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Zinc Finger Protein 638 (ZNF638).

Application

Quality Control Testing
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ZNF638 in NIH3T3 cell lysate.Tested applicationsWestern Blotting Analysis: A 1:1,000 dilution from a representative lot detected ZNF638 in PC12 cell lysate.Affinity Binding Assay: A representative lot of this antibody bound ZNF638 peptide with a KD of 4.1 x 10-7 in an affinity binding assay.Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ZNF638 in human colon tissue sections.Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ZNF638 in HeLa, A431, HepG2, and NIH 3T3 cells.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ZNF638, clone 4C19 ZooMAb, Cat. No. ZRB1186, is a recombinant Rabbit monoclonal antibody that targets ZNF638 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Zinc finger protein 638 (UniProt: Q14966; also known as Cutaneous T-cell lymphoma-associated antigen se33-1, CTCL-associated antigen se33-1, Nuclear protein 220, Zinc finger matrin-like protein, ZNF638) is encoded by the ZNF638 (also known as NP220, ZFML) gene (Gene ID: 27332) in human. ZNF638 is a nucleocytoplasmic transcription factor that binds to cytidine clusters in double-stranded DNA and plays a key role in the silencing of unintegrated retroviral DNA. It is also shown to act as an early regulator of adipogenesis by acting as a transcription cofactor of CEBPs, controlling the expression of PPAR and other pro-adipogenic genes, such as SREBF1. ZNF638 is selectively expressed in mature thermogenic adipocytes and tissues and its induction in response to classic stimuli promotes heat generation. It is shown to be upregulated by cAMP modulators in vitro and by cold exposure and by pharmacological stimulation of β-adrenergic signaling in vivo. Ectopic expression of ZNF638 is reported to increase adipogenesis in vitro, whereas its knockdown inhibits differentiation and decreases the expression of adipocyte-specific genes. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Perie, L., et al. (2019). J. Endocrine Soc. 3(12); 2326-2340; Meruvu, S., et al. (2011). J. Biol. Chem. 286(30); 26516-26523).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does this Anti-ZNF638 Antibody, clone 4C19 ZooMAb® Rabbit Monoclonal, works for Immunoprecipitation? Could you please send me the sequence that it targets to check whether it capture my ZNF638 isoform of interest or not? Thank you

    1 answer
    1. Anti-ZNF638 Antibody, clone 4C19 ZooMAb® Rabbit Monoclonal, is suitable for Affinity Binding. A representative lot of this antibody bound ZNF638 peptide with a KD of 4.1 x 10-7 in an affinity binding assay. This antibody specifically detects Zinc Finger Protein 638 (ZNF638) and targets an epitope within 21 amino acids from the C-terminal region. Please see the sequence as reported by UniProt accession number Q14966 https://www.uniprot.org/uniprotkb/Q14966/entry#sequences.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service