Skip to Content
Merck
All Photos(9)

Documents

ZRB1031

Sigma-Aldrich

Anti-Siglec-9 Antibody, clone 3F22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F22, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 50.08 kDa
observed mol wt ~670 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3F22 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Siglec-9. It targets an epitope within 15 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal, cytoplasmic domain of human Siglec-9.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Siglec-9 in Jurkat cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Siglec-9 in HepG2 and Raw264.7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Siglec-9 in human spleen tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected Siglec-9 in one million Human peripheral blood monouclear cells (PBMC).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Siglec-9, clone 3F22 ZooMAb®, Cat. No. ZRB1031, is a recombinant Rabbit monoclonal antibody that detects Siglec-9 and is tested for se in Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Sialic acid-binding Ig-like lectin 9 (UniProt: Q9Y336; also known as Siglec-9, CDw329, Protein FOAP-9, CD329) is encoded by the SIGLEC9 (also known as UNQ668/PRO1302) gene (Gene ID: 27180) in human. Sialic-acid-binding immunoglobulin-like lectins (Siglecs) belong to a lectin family of surface receptors that are predominantly expressed on hematopoietic cells with cell type and differentiation specificity. By binding with sialic acids at the terminal position of glycoproteins and lipids they regulate the intensity and duration of innate or adaptive immune response. Siglec-9 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 18-348), a transmembrane domain (aa 349-369), and a cytoplasmic domain (aa 370-463). It has one Ig-like V-type domain (aa 20-140) and two Ig-like C2-type domains (aa 146-229 and 236-336). It also contains one ITIM motif (aa 431-436) that is involved in modulation of cellular responses. The phosphorylated ITIM motif can bind the SH2 domain of several SH2-continaing phosphatases. It is expressed by peripheral blood leukocytes and is also found in a variety of other tissues at lower levels. Siglec9 serves as a putative adhesion molecule that mediate sialic-acid dependent binding to cells. It preferentially binds to alpha-2,3- or alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. Reduced expression of Siglec-9 on natural killer cells has been linked to persistent hepatitis B virus replication. Increased expression of soluble Siglec-9 has been reported in subjects with chronic obstructive pulmonary disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhao, D., et al. (2018). Front. Immunol. 9: 1124; Zeng, Z., et al. (2017). Sci. Rep. 7; Article 10116).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service