Skip to Content
Merck
All Photos(11)

Documents

ZMS1124

Sigma-Aldrich

Anti-m6A Antibody, clone 17-3-4-1 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

m6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

17-3-4-1, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG1λ

epitope sequence

Unknown

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 17-3-4-1 is a ZooMAb® Mouse recombinant monoclonal antibody that specifically detects N6-methyladenosine in proteins.

Immunogen

BSA-conjugated N6-methyladenosine.

Application

Quality Control Testing

Evaluated by DOT blot analysis with RNA oligos with m6A modification.

Dot Blot Analysis (DB): A 1:1,000 dilution of this antibody detected N6-methyladenosine in RNA oligos with m6A modification.

Tested Applications

Flow Cytometry Analysis: Flow Cytometry Analysis (FC): 1 µg of this antibody detected m6A in U2OS cells treated with 10 M BrdU, 24-28 h prior to UV-C treatment.

Immunocytochemistry Analysis: Immunocytochemistry Analysis (ICC): A 1:100 Dilution of this antibody detected m6A in U2OS cells treated with 10 M BrdU, 24-28 h prior to UV-C treatment.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is one of the most abundant post-transcriptional, reversible modification of RNA. m6A is not only an RNA marker, but also a genomic DNA marker (m6dA) that is detected in prokaryotes and lower eukaryotes. In higher eukaryotes, m6dA levels are abundant during embryogenesis and significantly decrease in adult tissues. It is a developmentally regulated RNA modification that is dynamically modified. The m6A modification is an important physiological regulator involved in cell renewal and differentiation and may function in post-transcriptional fine-tuning of gene expression and may provide a faster means of maximizing gene expression. It is shown to exhibit tissue-specific regulation and is reported to be elevated throughout brain development. The methylation of adenosine is catalyzed by a multicomponent complex that is composed of METTL3/MT-A70, METTL14, and WTAP in mammals. METTL3 and METTL14 are responsible for the methyltransferase activity of the complex, and WTAP mediates substrate recruitment. Fat mass and obesity-associated (FTO) and ALKBH5 are two identified demethylases that are reported to catalyze oxidative de-methylation of m6A. These enzymes work in a cooperative manner to maintain the balance of intracellular m6A levels. It has been shown that m6A writers that consist of the methyltransferase complex catalyze this modification positively and it is reversed by m6A erasers with demethylase activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Meyer, KD., et al. (2012). Cell. 149(7):1635-1646).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service