Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1096

Sigma-Aldrich

Anti-Sulfotyrosine Antibody, clone Sulfo-1C-A2 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Sulfo-1C-A2, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 55 kDa
observed mol wt ~55 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone Sulfo-1C-A2 is a ZooMAb® Mouse recombinant monoclonal antibody that specifically detects sulfated tyrosine residues on various proteins. It does not recognize phosphotyrosine or unmodified tyrosines.

Immunogen

Developed using a phage display library as described in Kehoe, JW., et al. (2006). Mol. Cell. Proteomics. 5(12); 2350-2563. The selection antigens were KAKISDP-DY(SO3)MTGYMDAC and KDKKYATEY(SO3)-EYLDYDFC.

Application

Quality Control Testing

Evaluated by Western Blotting with Bovine Fibrinogen.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Sulfotyrosine in Bovine Fibrinogen, but not in Fibrinogen treated with Sodium Acetate and Abalone Sulfatase.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Sulfotyrosine in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lo detected Sulfotyrosine in Human umbilical vein endothelial cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Protein tyrosine O-sulfation is a post-translational modification mediated by two Golgi tyrosylprotein sulfotransferases (TPST1 and TPST2) that catalyze the transfer of sulfate to tyrosine residues in secreted and transmembrane proteins. These enzymes catalyze the transfer of sulfate from 3 -phosphoadenosine 5 -phosphosulfate to tyrosine residues in proteins. TPST 1 and 2 are type II transmembrane proteins that reside in the trans-Golgi network and have luminally oriented catalytic domain. Hence, tyrosine sulfation occurs only on soluble and transmembrane proteins that transit the Golgi en route to either secretion or incorporation into the plasma membrane. Tyrosine sulfation occurs in all multicellular organisms and up to 1% of the tyrosine residues in the total protein content of a cell can be sulfated. Tyrosine sulfation is shown to be essential for several biological processes, such as optimal proteolytic processing of progastrin, proteolytic activation of coagulation factors V and VIII by thrombin, proteolysis of the complement C4a-chain by C1s. Sulfotyrosine is also reported to be involved in the entry of several parasites and viruses into cells. Tyrosine sulfation of the HIV-1 co-receptor CCR5 is required for viral entry into host cells. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Huang, C., et al. (2007). Science. 317(5846); 1930-1934; Kehoe, JW., et al. (2006). Mol. Cell. Proteomics. 5(12); 2350-2563; Hoffhines, AJ., et al. (2006). J. Biol. Chem. 281(49); 37877-37887).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service