Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1054

Sigma-Aldrich

Anti-Centrin Antibody, clone 20H5 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

20H5, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~19.46 kDa
observed mol wt ~18 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

Chlamydomonas reinhardtii, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 20H5 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects centrin (Caltractin). It targets an epitope within the C-terminal region.

Immunogen

Recombinant Chalamydomonas reinhardtii centrin.

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse testis tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Centrin in Mouse testis tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Centrin in human testis tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Centrin in human testis tissue sections.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Centrin in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Centrin, clone 20H5 ZooMAb®, Cat. No. ZMS1054, is a recombinant Mouse monoclonal antibody that specifically targets Centrin and is tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Caltractin (UniProt: P05434; also known as 20 kDa calcium-binding protein, Centrin) is encoded by the VFL2 gene (Gene ID: 5725094) in Chlamydomonas reinhardtii (algae). Centrin is a highly conserved calmodulin-like calcium-binding phospho-protein that contain four EF hands and are associated with centrosome-related structures. In Chlamydomonas reinhardtii, centrin is shown to play an essential role in microtubule severing during flagellar excision during which time stellate fibers of the transition zone contract and displace the microtubule doublets of the axoneme inward. The resulting shear force and torsional load generated during inward displacement leads to microtubule severing immediately distal to the central cylinder of the transition zone. Centrin is also present is all eukaryotic cells and contain four helix-loop-helix motifs that all conform to the EF-hand consensus for Ca2+ binding. Two of these motifs exhibit high affinity for Ca2+ two display lower affinity. Stability of centrin is regulated by phosphorylation of serine residue in the C-terminal region by aurora kinase A, which also localizes with centrin at the centrosome. Their levels are shown to be the highest from prophase to metaphase stage followed by precipitous loss in the late stage of mitosis. In human breast cancers higher levels of aurora kinase A and phospho-centrin have been observed. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sanders, MA., and Salisbury, JL. (1994). J. Cell Biol. 24(5); 795-805; Lutz, W., et al. (2001). J. Biol. Chem. 276(23); 20774-20780; Lukasiewicz, KB., et al. (2011). PLoS ONE 6(6); e21291).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service