Z757772
Sartorius filter pipette tips
volume range 0.5-20 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Synonym(s):
filter pipette tips, pipetting tips, pipettor tips
Select a Size
€186.00
Select a Size
About This Item
RNase free
barrier
filter
€186.00
Recommended Products
material
colorless
polypropylene (Virgin)
sterility
sterile
feature
DNase free
RNase free
barrier
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 790021F
maximum volume
20 μL
volume range
0.5-20 μL
volume
0.5—20 μL
binding type
low binding surface
Related Categories
General description
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
- Pipette foaming liquids like buffers and proteins
- Pipette solvents
- Reverse pipetting
- Multiple dispensing functions of electronic pipettes
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)