Z744291
Wilmad® 4 mm ZrO2 Open Ended Rotor for Bruker® MAS Probe
zirconia, Laser marked, pack of 1 ea
Synonym(s):
Solid State NMR, MAS NMR, Solid State NMR, MAS NMR,
About This Item
AC/DC input
18 kHz
material
zirconia
description
Rotor Body laser marked with a unique serial number and permanent tachometer mark on the base.
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WP-501-4181
parameter
at 100-200 °C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- No need to solvate solid samples
- Homogeneous ceramic material structure
- Strength 1,000 Mpa
- Finish 0.05 µ mirror gloss
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service