Key Documents
Z744144
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
pack of 1
Synonym(s):
EPR cryo dewar, EPR dewar
About This Item
UNSPSC Code:
41121804
packaging
pack of 1
manufacturer/tradename
Wilmad WG-816-Q
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Legal Information
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)