This product is not sterile. In general, polypropylene may be autoclaved. Alternatively, the item may be placed in the hood under UV or dissinfected using 70% Ethanol.
Z744074
Reagent Reservoir with Hinged-Lid
reservoir capacity (175 mL), polypropylene
Select a Size
€39.20
Estimated to ship on29 May 2025
Select a Size
About This Item
€39.20
Estimated to ship on29 May 2025
Recommended Products
material
polypropylene
feature
lid
packaging
case of 10 ea
manufacturer/tradename
Excel Scientific RSV-L175-N10PK
reservoir capacity
(175 mL)
General description
- Volume graduations to 150 mL
- A reinforced base that won′t wobble on the counter, preventing spills
- A new easy-to-open latch that can be manipulated with one hand
- Greater clarity for improved visibility of contents
- Reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved.
- The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste.
- Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents.
- With lids closed, reagent reservoirs are stackable.
- A perimeter ridge on the lid prevents sliding.
- Construction is clear, chemical-resistant, autoclavable polypropylene.
- Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is this product sterile?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service