Skip to Content
Merck
All Photos(1)

Key Documents

Z742633

Tube-O-DIALYZER mini dialysis system

Medi, MWCO 50 K

Synonym(s):

semi-permeable membrane tube, single use tube dialysis

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€243.00

€243.00


Estimated to ship on17 April 2025



Select a Size

Change View
20 EA
€243.00

About This Item

UNSPSC Code:
41123100
NACRES:
NB.24

€243.00


Estimated to ship on17 April 2025


feature

Medi

packaging

pkg of 20 ea

manufacturer/tradename

G-BioSciences 786-619

pore size

50 K MWCO

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dialysis is a popular technique used for the exchange of buffer medium across semi-permeable membranes. Dialysis devices are available in many configurations for research applications. We offer innovative dialysis devices and accessories for processing small samples. The Tube-O-DIALYZER mini dialysis system is designed for small sample volumes of 20μl to 2.5ml.
The Tube-O-DIALYZER tube comes in two ideal sizes; the Micro unit allows efficient dialysis of 20-250μl samples and the Medi unit is optimized for samples between 0.2-2.5ml. Both sizes are available with membranes with molecular weight cutoffs (MWCO) of 1k, 4k, 8k, 15k, and 50k Daltons.
Tube-O-DIALYZER comes in packs of 20, each supplied with 6 appropriately sized floats and storage caps to allow storage of dialyzed samples. For added convenience, Tube-O-DIALYZER is also available as a mixed kit containing 10 Micro and 10 Medi units.

Application

  • Dialysis of small sample volumes
  • Equilibrium dialysis for buffer exchange
  • Dialysis for single use applications, such as infectious and radioactive samples

Legal Information

Tube-O-DIALYZER is a trademark of Geno Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Will organic solution like ethanol cause potential damage to this dialysis membrane?

    1 answer
    1. Alcohols are typically compatible with this product. Strong acids and bases are not recommended. See the image below to review the product compatibility table:

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service