Key Documents
Z742401
PELCO® Sliding Latch Microslide Storage Boxes
blue, Holds 50 x slides 76x26 mm
Synonym(s):
slide storage
Select a Size
12 EA
€163.00
€163.00
Check Cart for Availability
Select a Size
Change View
12 EA
€163.00
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
€163.00
Check Cart for Availability
material
acrylonitrile-butadiene-styrene (ABS)
blue
feature
Holds 50 x slides 76x26 mm
packaging
pack of 12 ea
manufacturer/tradename
Ted Pella 2304-4-CS
L × W × H
164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.)
General description
Exceptional slide storage containers providing maximum protection for your microscope slides.
Features and Benefits
  • Numbered index sheet in lid for quick identification
  • Foam liner
  • Stackable
  • ABS-durable/impact resistant
  • Hinged lid
  • Unique sliding compression latch
Legal Information
PELCO is a registered trademark of Ted Pella, Inc
Choose from one of the most recent versions:
Certificates of Analysis (COA)