Skip to Content
Merck
All Photos(1)

Key Documents

Z740415

SecurTainer III specimen containers

size 90 mL

Sign Into View Organizational & Contract Pricing

Select a Size

400 EA
€326.00

€326.00


Estimated to ship on28 May 2025



Select a Size

Change View
400 EA
€326.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

€326.00


Estimated to ship on28 May 2025


material

polyethylene cap
polypropylene container

packaging

pack of 400 ea

manufacturer/tradename

Simport C577-90W

size

90 mL

Looking for similar products? Visit Product Comparison Guide

General description

Chemically resistant and shatterproof, The SecurTainer III Specimen Containers are supplied with a leakproof screw cap, particularly important when transporting hazardous material. Ideal for transport and storage of urine, sputum and most liquids or particulate samples.

Containers are uniquely stackable, shatter resistant and are manufactured from virgin, high-clarity polypropylene. Available in many sizes from 20 to 120 ml. Containers and caps packaged separately in bags of 100. Non sterile. Functional temperature range: -90 °C to +100 °C.

Legal Information

SecurTainer is a trademark of Simport

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service