Key Documents
Z740376
Eco-Friendly SpecTainer, tamper evident, biodegradable
size 60 mL, non-sterile
Select a Size
500 EA
€563.00
€563.00
Estimated to ship on24 May 2025
Select a Size
Change View
500 EA
€563.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
€563.00
Estimated to ship on24 May 2025
material
polyethylene cap
polypropylene container
sterility
non-sterile
packaging
pack of 500 ea
manufacturer/tradename
Simport C566-60DOECO
size
60 mL
General description
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
Legal Information
SpecTainer is a trademark of Simport
Choose from one of the most recent versions:
Certificates of Analysis (COA)