Yes, product Z740367-40EA Urisafe® urine collection container, is CE marked .
Z740367
Urisafe® urine collection container
size 24 Hour, volume 3 L
Select a Size
€248.00
Estimated to ship on12 April 2025
Select a Size
About This Item
€248.00
Estimated to ship on12 April 2025
Recommended Products
material
high-density polyethylene container
polypropylene cap
packaging
pack of 40 ea
manufacturer/tradename
Simport B350
size
24 Hour
volume
3 L
3000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Snap valve pouring spout incorporated
- Leakproof: a cap liner ensures safe sample transport
The unique snap valve pour spout:
- Easily popped open or pushed shut
- Offers dripless pouring
- Controlled flow rate for better handling of poured volumes
- Reduced risks of aerosol contamination when pouring
- Eliminates splashing and exposure to hazardous body fluids
Low form design:
- Convenient refrigerator storage with less wasted space
- Anatomically designed for ease of patient use
Large central handle:
- Can be gripped comfortably with three fingers
- Permits pouring off of samples with ease and reduces fatigue
Chemically resistant:
- Metal, latex, zinc and fluorescence free
- Resistant to hydrochloric acid.
- Can be subjected to freezing, thawing and EtO gas sterilisation without causing changes in materials or physical appearance
Graduated vertically and horizontally:
- Graduations are easy to read
- Place container upright and read in 100 ml increments
- Can be gamma sterilized
1 The unique snap valve pour spout is easily popped open and offers dripless pouring
2 Specially designed leakproof screw cap with liner for safer transport
3 Large centrally located handle for good balance and ease of manipulation
4 Metal free and resistant to hydrochloric acid
5 Sample volumes can be read in 100 ml increments
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Please confirm whether product Z740367-40EA has a CE mark.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service