Skip to Content
Merck
All Photos(3)

Key Documents

Z726931

Spectra-Por® Float-A-Lyzer® G2

red, 10 mL, MWCO 20 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
€193.00

€193.00


Check Cart for Availability


Select a Size

Change View
12 EA
€193.00

About This Item

UNSPSC Code:
41123100
NACRES:
NB.22

€193.00


Check Cart for Availability

material

red

packaging

pack of 12 ea (dry with glycerol)

manufacturer/tradename

Spectrum Labs G235069

pore size

20 kDa MWCO

capacity

10 mL

Looking for similar products? Visit Product Comparison Guide

General description


  • 95 - 98% Sample Recovery
  • Highest Membrane Purity
  • Superior Handling & Leak Prevention
  • Volume Specific Dilution Control
  • Complete & Ready-to-Use Packaging

Spectrum′s proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 4 precise MWCO′s (color coded for MWCO variants) with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval. Only the Float-A-Lyzer® G2 assures a 95-98% sample recovery while maintaining 99% sample purity and <1% sample dilution.

The leak-proof screw-on cap with sealing o-ring provides easy access with included pipette for loading, in-process testing and sample retrieval, without the risk of needle punctures. The included floatation ring improves sample buoyancy and vertical orientation during dialysis. The sleek design allows multiple samples to be dialyzed in the same reservoir.
No additional kits, buoys or accessories are required. The Float-A-Lyzer G2 is packaged dry and ready-to-go with all components included. Simply rinse with water, load the sample with a pipette, and dialyze.
Spectrum Laboratories, Inc. introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation of Ready-to-Use laboratory dialysis devices featuring a Ultra-pure Biotech Cellulose Ester (CE) Membrane

Legal Information

Float-A-Lyzer is a registered trademark of Spectrum Laboratories, Inc.
Spectra/Por is a registered trademark of Spectrum Laboratories, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service