Key Documents
Z711039
ART® non-filtered tip
HLT 200G, size 20-200 μL, Pure packed, sterile
Synonym(s):
pipet tips, pipette tips
Select a Size
1 PKG
€105.00
5 PKG
€388.00
€105.00
Estimated to ship on24 May 2025
Select a Size
Change View
1 PKG
€105.00
5 PKG
€388.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
20—200 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier: no
Pure packed
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 3531
€105.00
Estimated to ship on24 May 2025
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier: no
Pure packed
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Molecular BioProducts 3531
maximum volume
200 μL
size
20-200 μL
volume
20—200 μL
General description
Genomic tip.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)